Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
HDPE plastic bottle
Catalog number 401130250
Price (USD)/ Each
25.65
Online exclusive
27.80 
Save 2.15 (8%)
-
Add to cart
Quantity:
25 g
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
27.80 
Save 2.15 (8%)
Add to cart
Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Catalog number401130250
Price (USD)/ Each
25.65
Online exclusive
27.80 
Save 2.15 (8%)
-
Add to cart
Chemical Identifiers
CAS506-87-6
IUPAC Namecarbonic acid diamine
Molecular FormulaCH8N2O3
InChI KeyPRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILESN.N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals and/or chunks
Titration excess HCl>=30.0 % (NH3)
Heavy metals (ICP-OES)=<5 ppm
Non-volatile matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only