Applied Biosystems™

Beta Cyanoethyl Phosphoramidite, dA(Bz)

Catalog number: 401159
Have Questions?
Applied Biosystems™

Beta Cyanoethyl Phosphoramidite, dA(Bz)

Catalog number: 401159
Have Questions?
Catalog Number
401159
Unit Size
2 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4011592 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The dA(Bz) Beta Cyanoethyl Phosphoramidite is used as a component of a deprotecting chemistry set including dC(Bz), dG(iBu) and T. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.

β-cyanoethyl phosphoramidites are for use on ABI synthesizers to guarantee you high quality nucleic acid synthesis.

• Proprietary manufacturing processes yield high-purity β-cyanoethyl phosphoramidites, which in turn guarantee production of high-quality DNA at all scales.
• β-cyanoethyl phosphoramidites are use-tested and optimized for use on the ABI synthesizers to guarantee you high-quality nucleic acid synthesis.

High Quality
Our phosphoramidites benefit from proprietary manufacturing processes that yield high-purity products. In synthesis they perform at a correspondingly high level, producing pure, high quality DNA at all scales. We guarantee that 5'-dimethoxytrityl nucleoside β-cyanoethyl phosphoramidites, monomer intermediates for licensed β-cyanoethyl phosphoramidite chemistry, will produce DNA that is at least 99.8% chemically authentic when used with recommended synthesis cycles.

Dry phosphoroamidites (powder form) are guaranteed for one year from the date of shipment. Lifetime of diluted phosphoramidites in residence on the instrument is two weeks when used with CPG synthesis supports, and three weeks when used with polystyrene supports.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
3900 DNA Synthesizer
Product Type
dA(Bz) Beta CEP
Nucleoside Type
Deoxyadenosine (dA)
Shipping Condition
Room Temperature
Reagent Type
Phosphoramidite
Form
Powder

Contents & Storage

Store in Freezer (-5°C to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text