Ammonium persulfate, 98+%, Thermo Scientific Chemicals
Ammonium persulfate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium persulfate, 98+%, Thermo Scientific Chemicals

Oxidation reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
500 g
2 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 401161000
Price (USD)/ Each
58.65
Online exclusive
64.80 
Save 6.15 (9%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
64.80 
Save 6.15 (9%)
Add to cart
Ammonium persulfate, 98+%, Thermo Scientific Chemicals
Catalog number401161000
Price (USD)/ Each
58.65
Online exclusive
64.80 
Save 6.15 (9%)
-
Add to cart
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration with KMnO4>=98.0 %
Heavy metals (as Pb)=<0.005 %
Residue after ignition=<0.05 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only