Thermo Scientific Chemicals

4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade

Catalog number: 401261000
100 g, Each
Thermo Scientific Chemicals

4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade

Catalog number: 401261000
100 g, Each
Quantity
Catalog number: 401261000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-73-4
IUPAC Name
sodium 4-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular Formula
C18H14N3NaO3S
InChI Key
MLVYOYVMOZFHIU-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
Appearance (Color)
Ochre to orange
Appearance (Form)
Powder
Infrared spectrum
Conforms
Visual transition interval
from pH 1.4 (red) to pH 2.6 (yellow)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text