Thermo Scientific Chemicals

L(+)-Ascorbic acid, ACS reagent, Thermo Scientific Chemicals

Catalog number: 401475000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

L(+)-Ascorbic acid, ACS reagent, Thermo Scientific Chemicals

Catalog number: 401475000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 401475000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-81-7
IUPAC Name
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Formula
C6H8O6
InChI Key
CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Titration Iodimetric
>=99.0 %
Heavy metals (ICP-OES)
=<20 ppm
Specific optical rotation
+20.5° to +21.5° (25°C, 589 nm) (c=10, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text