L(+)-Asparagine monohydrate, Thermo Scientific Chemicals
L(+)-Asparagine monohydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(+)-Asparagine monohydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Catalog number 401482500
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS5794-13-8
IUPAC Name(2S)-2-amino-3-carbamoylpropanoic acid hydrate
Molecular FormulaC4H10N2O4
InChI KeyRBMGJIZCEWRQES-DKWTVANSSA-N
SMILESO.N[C@@H](CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Chlorides=<200 ppm
Titration with HClO499.0 to 101.0 % (on dried substance)
Loss on drying10.5 % to 12.5 % (1 g, 130°C, 3 h)
Sulfated ash=<0.1 % (1 g)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only