Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminon, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 401500250
Price (USD)/ Each
45.65
Online exclusive
50.00 
Save 4.35 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.00 
Save 4.35 (9%)
Add to cart
Aluminon, ACS reagent, Thermo Scientific Chemicals
Catalog number401500250
Price (USD)/ Each
45.65
Online exclusive
50.00 
Save 4.35 (9%)
-
Add to cart
Chemical Identifiers
CAS569-58-4
IUPAC Nametriammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular FormulaC22H25N3O9
InChI KeyUDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.2 %
Appearance (Color)Red to yellow-brown to red-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Sensitivity(to Aluminium) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only