Thermo Scientific Chemicals

Azelaic acid, 98%, Thermo Scientific Chemicals

Catalog number: 401521000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Azelaic acid, 98%, Thermo Scientific Chemicals

Catalog number: 401521000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 401521000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-99-9
IUPAC Name
nonanedioic acid
Molecular Formula
C9H16O4
InChI Key
BDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCC(O)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text