Chemical Identifiers
IUPAC Namehydrogen 2-[2-(1-carbamimidoyl-1-methylethyl)diazen-1-yl]-2-methylpropanimidamide chloride
Molecular FormulaC8H19ClN6
InChI KeyQMYCJCOPYOPWTI-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(C)(N=NC(C)(C)C(N)=N)C(N)=N
Molecular Weight (g/mol)234.73
View more