Thermo Scientific Chemicals

2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 401560500
50 g, Plastic Bottle, Each
Thermo Scientific Chemicals

2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 401560500
50 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 401560500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2997-92-4
IUPAC Name
hydrogen 2-[2-(1-carbamimidoyl-1-methylethyl)diazen-1-yl]-2-methylpropanimidamide chloride
Molecular Formula
C8H19ClN6
InChI Key
QMYCJCOPYOPWTI-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC(C)(N=NC(C)(C)C(N)=N)C(N)=N
Appearance (Color)
White to off-white
Appearance (Form)
Granules
Infrared spectrum
Conforms
Titration Argentometric
97.5 to 102.5 % (on dry substance)
Loss on drying
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text