Thermo Scientific Chemicals

Barium chloranilate trihydrate, Thermo Scientific Chemicals

Catalog number: 401661000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Barium chloranilate trihydrate, Thermo Scientific Chemicals

Catalog number: 401661000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 401661000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
32458-20-1
IUPAC Name
2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione barium trihydrate
Molecular Formula
C6H8BaCl2O7
InChI Key
JFBLKYVZQCFPJE-UHFFFAOYSA-N
SMILES
O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
Appearance (Color)
Brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Suitability
(for Sulfate determination) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text