Thermo Scientific Chemicals

Barium diphenylaminesulfonate, pure, indicator grade

Catalog number: 401680050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Barium diphenylaminesulfonate, pure, indicator grade

Catalog number: 401680050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 401680050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6211-24-1
IUPAC Name
barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
Molecular Formula
C24H20BaN2O6S2
InChI Key
IVCNVXFNTKXMCA-UHFFFAOYSA-L
SMILES
[Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Appearance (Color)
Off-white to grey
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Loss on drying
=<1 % (105°C)
Sensitivity
(as indicator) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text