Chemical Identifiers
IUPAC Namebarium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
Molecular FormulaC24H20BaN2O6S2
InChI KeyIVCNVXFNTKXMCA-UHFFFAOYSA-L
SMILES[Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Molecular Weight (g/mol)633.88
View more