Chemical Identifiers
CAS632-99-5
IUPAC Namehydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular FormulaC20H20ClN3
InChI KeyAXDJCCTWPBKUKL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
View more