Thermo Scientific Chemicals

Basic Fuchsin, 70+%, pure, Thermo Scientific Chemicals

Catalog number: 401695000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Basic Fuchsin, 70+%, pure, Thermo Scientific Chemicals

Catalog number: 401695000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 401695000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
632-99-5
IUPAC Name
hydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular Formula
C20H20ClN3
InChI Key
AXDJCCTWPBKUKL-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Dye content
>=70.0 %
UV Lambda max
547 to 552 nm (in 50% ethanol)
Absorption Ratio P-15/P+15
1.16 to 1.35
Appearance (Color)
Dark green
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text