Thermo Scientific Chemicals

Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 401790250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 401790250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 401790250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1670-14-0
IUPAC Name
hydrogen benzenecarboximidamide chloride
Molecular Formula
C7H9ClN2
InChI Key
LZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(=N)C1=CC=CC=C1
Appearance (Color)
White to off-white
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
HPLC
>=98.5 %
Water
=<1.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text