Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 401791000
Price (USD)/ Each
393.65
Online exclusive
437.00 
Save 43.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
393.65
Online exclusive
437.00 
Save 43.35 (10%)
Add to cart
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Catalog number401791000
Price (USD)/ Each
393.65
Online exclusive
437.00 
Save 43.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1670-14-0
IUPAC Namehydrogen benzenecarboximidamide chloride
Molecular FormulaC7H9ClN2
InChI KeyLZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(=N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=98.5 %
Water=<1.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only