Thermo Scientific Chemicals

Benzenesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 401850250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzenesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 401850250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 401850250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
515-42-4
IUPAC Name
sodium benzenesulfonate
Molecular Formula
C6H5NaO3S
InChI Key
MZSDGDXXBZSFTG-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)C1=CC=CC=C1
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=97.5 %
Loss on drying
=<2 % (1 g, 105°C, 3 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text