Benzenesulfonic acid, sodium salt, 98%
Benzenesulfonic acid, sodium salt, 98%
Benzenesulfonic acid, sodium salt, 98%
Benzenesulfonic acid, sodium salt, 98%
Thermo Scientific Chemicals

Benzenesulfonic acid, sodium salt, 98%

CAS: 515-42-4 | C6H5NaO3S | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 401851000
Price (USD)
48.65
Special offer
Online exclusive
Ends: 26-Sep-2025
60.30
Save 11.65 (19%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 26-Sep-2025
60.30
Save 11.65 (19%)
Each
Add to cart
Chemical Identifiers
CAS515-42-4
IUPAC Namesodium benzenesulfonate
Molecular FormulaC6H5NaO3S
InChI KeyMZSDGDXXBZSFTG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<2 % (1 g, 105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link