Thermo Scientific Chemicals

Benzoyl chloride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 402030025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Benzoyl chloride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 402030025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 402030025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-88-4
IUPAC Name
benzoyl chloride
Molecular Formula
C7H5ClO
InChI Key
PASDCCFISLVPSO-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC=C1
Titration excess NaOH
98.0 to 100.5 %
Heavy metals (as Pb)
=<10 ppm
Refractive index
1.5520 to 1.5540 (20°C, 589 nm)
Residue after ignition
=<0.005 %
Phosphorous compounds (as P)
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text