Thermo Scientific Chemicals

Leucomalachite Green

Catalog number: 402110250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Leucomalachite Green

Catalog number: 402110250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 402110250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
129-73-7
IUPAC Name
4-{[4-(dimethylamino)phenyl](phenyl)methyl}-N,N-dimethylaniline
Molecular Formula
C23H26N2
InChI Key
WZKXBGJNNCGHIC-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
Appearance (Color)
White to light green to light brown to light blue
Appearance (Form)
Powder
Infrared spectrum
Conforms
UV spectrum
Complies

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text