Thermo Scientific Chemicals

Betaine monohydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 402191000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Betaine monohydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 402191000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 402191000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-47-6
Appearance (Color)
White
Titration with HClO4
99.0 to 101.0 % (On dry substance)
Loss on drying
11 % to 15 % (140°C, 3 h)
Sulfated ash
=<0.1 %
Heavy metals (as Pb)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text