Bicyclohexyl, 99%, Thermo Scientific Chemicals
Bicyclohexyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bicyclohexyl, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 402211000
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
336.00 
Save 50.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
336.00 
Save 50.35 (15%)
Add to cart
Bicyclohexyl, 99%, Thermo Scientific Chemicals
Catalog number402211000
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
336.00 
Save 50.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,1'-bi(cyclohexane)
Molecular FormulaC12H22
InChI KeyWVIIMZNLDWSIRH-UHFFFAOYSA-N
SMILESC1CCC(CC1)C1CCCCC1
Molecular Weight (g/mol)166.31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4780 to 1.4800 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only