Thermo Scientific Chemicals

Biebrich Scarlet, dye content approx. 70%

Catalog number: 402221000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Biebrich Scarlet, dye content approx. 70%

Catalog number: 402221000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 402221000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4196-99-0
IUPAC Name
disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
Molecular Formula
C22H14N4Na2O7S2
InChI Key
YCKMEQRBEVVZQF-VKZXTWAVSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Dye content
>=65 %
Loss on drying
=<15 % (110°C)
Appearance (Color)
Red to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text