Thermo Scientific Chemicals

N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 97%, Thermo Scientific Chemicals

Catalog number: 402280050
5 g, Each
Thermo Scientific Chemicals

N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 97%, Thermo Scientific Chemicals

Catalog number: 402280050
5 g, Each
Quantity
Catalog number: 402280050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4605-14-5
IUPAC Name
(3-azaniumylpropyl)({3-[(3-azaniumylpropyl)azaniumyl]propyl})azanium
Molecular Formula
C9H28N4
InChI Key
ZAXCZCOUDLENMH-UHFFFAOYSA-R
SMILES
[NH3+]CCC[NH2+]CCC[NH2+]CCC[NH3+]
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
GC
>=92 %
Appearance (Form)
Liquid
Refractive index
1.4900 to 1.4930 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text