Thermo Scientific Chemicals

2,2'-Bithiophene, 97%, Thermo Scientific Chemicals

Catalog number: 402690025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2'-Bithiophene, 97%, Thermo Scientific Chemicals

Catalog number: 402690025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 402690025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
492-97-7
IUPAC Name
2,2'-bithiophene
Molecular Formula
C8H6S2
InChI Key
OHZAHWOAMVVGEL-UHFFFAOYSA-N
SMILES
S1C=CC=C1C1=CC=CS1
Appearance (Color)
Off-white to dark green to blue
Appearance (Form)
Crystals or low melting crystalline mass
Infrared spectrum
Conforms
GC
>=96.0 %
Melting point
30°C to 35°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text