Thermo Scientific Chemicals

Borane-trimethylamine complex, 97%, Thermo Scientific Chemicals

Catalog number: 402741000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Borane-trimethylamine complex, 97%, Thermo Scientific Chemicals

Catalog number: 402741000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 402741000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-22-9
IUPAC Name
trimethylamine borane
Molecular Formula
C3H12BN
InChI Key
WVMHLYQJPRXKLC-UHFFFAOYSA-N
SMILES
B.CN(C)C
Appearance (Color)
White
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration with Na2S2O3
>=96.0 %
Melting point
92°C to 96°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text