Applied Biosystems™

TFA Aminolink Phosphoramidite, 0.25 g

Catalog number: 402872
Have Questions?
Applied Biosystems™

TFA Aminolink Phosphoramidite, 0.25 g

Catalog number: 402872
Have Questions?
Catalog Number
402872
Unit Size
0.25 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4028720.25 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Each contains 0.25 g TFA Aminolink.

TFA Aminolink is used for 5’ attachment of amine group during oligonucleotide synthesis.

We offer several phosphoramidites that impart linkers for coupling other labels, and the means to add a phosphate during oligonucleotide synthesis. The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.

• Specialty phosphoramidites allow online addition of labels and linkers during synthesis, saving time in production of labeled probes and primers.
• They are designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
• We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis.

Amine linker:
An oligonucleotide incorporating a primary amine readily accepts a variety of labels. TFA Aminolink™ phosphoramidite establishes that amino group at the 5'-end during nucleic acid synthesis.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Powder
For Use With (Equipment)
3900 DNA Synthesizer
Product Type
Aminolink CEP
Nucleoside Type
None (Modifier)
Shipping Condition
Dry Ice
Reagent Type
Phosphoramidite
Nucleic Acid Type
None (Modifier)
For Use With (Application)
ABI Synthesizers, Guaranteeing Reliable Nucleic Acid Synthesis

Contents & Storage

Store in a cool, dry place.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text