Thermo Scientific Chemicals

Bromocresol Green, sodium salt

Catalog number: 402940500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromocresol Green, sodium salt

Catalog number: 402940500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 402940500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67763-24-0
IUPAC Name
sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular Formula
C21H13Br4NaO5S
InChI Key
HEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
Infrared spectrum
Conforms
Appearance (Color)
Dark brown to green-black
Appearance (Form)
Powder
Clarity of solution
Passes test
Visual transition interval
from pH 3.8 (yellow) to pH 5.4 (blue)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text