Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps

Catalog number: 403012
Have Questions?
Applied Biosystems™

MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps

Catalog number: 403012
Have Questions?
Catalog Number
403012
Unit Size
20 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
40301220 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps is optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems™ 96-Well Real-Time PCR systems and thermal cyclers.

• Frosted to minimize interfering fluorescence from wells of cycling block
• Screened to eliminate auto-fluorescent plates
• Unique design provides a barrier to ambient air to help insure well-to-well temperature uniformity
• Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
• MicroAmp™ Optical 8-Cap Strips included

Oil-Free PCR Amplification
MicroAmp™ disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification. Proprietary MicroAmp™ plates are essential for high-performance PCR and help ensure the greatest temperature and amplification uniformity for your samples.

Specifications

Barcode
Includes Barcode
Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Color
Optical
Product Type
96-Well Reaction Plate
For Use With (Equipment)
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Volume (Metric) Well
0.3 mL
No. of Wells
96
For Use With (Application)
PCR
Quantity
20 Plates
Reaction Speed
Standard
Unit Size
20 plates

Contents & Storage

Contains 20 plates with 300 Optical 8-cap Strips
(equivalent to catalog # 4306737 and 4323032)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text