Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ 96-Well Tray/Retainer Set

Catalog number: 403081
Have Questions?
Applied Biosystems™

MicroAmp™ 96-Well Tray/Retainer Set

Catalog number: 403081
Have Questions?
Catalog Number
403081
Unit Size
10 sets
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
40308110 sets
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems™ MicroAmp™ Tray/Retainer Sets, available in a 96- or 24-well format, hold MicroAmp™ Reaction Tubes without Caps for processing on the Veriti™ 96-Well Thermal Cycler, 96-Well GeneAmp™ PCR System 9700, or the Applied Biosystems 2720 Thermal Cycler.

• Available in 96- or 24-well format, offering flexibility
• Designed for compatibility with MicroAmp™ Reaction Tubes and MicroAmp™ 8-Strip Reaction Tubes

Convenience and Performance
The MicroAmp™ Tray/Retainer Sets allow you to organize your tubes while adding samples and mixing. This handy tool reduces spillage and tube mix-ups by securing and ordering your tubes on your lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the GeneAmp Thermal Cycler. Not only does the tray eliminate the hassle of transferring the tubes but it also provides the thermal contact necessary for high-performance thermal cycling.


Note: See user's manual or package insert for limited label license, and trademark information.

Specifications

Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Color
White
Product Type
96-Well Tray/Retainer Set
For Use With (Equipment)
2720 Thermal Cycler, 7000 System, AB 2720 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
10 Set
Shipping Condition
Room Temperature

Contents & Storage

Holds up to 96 MicroAmp™ tubes w⁄o caps (0.2 mL) for processing either on the System 9600 or 9700. Use with MicroAmp™ 96-Well Base P⁄N N8010531

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text