Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Catalog number: 403083
Have Questions?
Applied Biosystems™

MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Catalog number: 403083
Have Questions?
Catalog Number
403083
Unit Size
20 assemblies
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 assemblies
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
40308320 assemblies
-
403086100 assemblies
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set is a completely pre-assembled unit containing MicroAmp™ Reaction Tubes without caps already loaded in a MicroAmp™ 96-Well Tray/Retainer Set. MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Sets are optimally designed for precise PCR on Applied Biosystems™ 0.2 mL thermal cyclers.

• Completely pre-assembled for convenience
• Available in economical 100-unit bulk packs
• Non-sterile and autoclavable

Greater Sample Uniformity
Use with all instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay. Proprietary MicroAmp™ Reaction Tubes are essential for high-performance PCR and ensure the greatest temperature and amplification uniformity for your samples. The MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set requires caps or full plate covers to seal the tubes.

Note: See user's manual or package insert for limited label license and trademark information.

Specifications

Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Color
Natural
Product Type
96-Well Reaction Tube/Tray/Retainer Set
For Use With (Equipment)
2720 Thermal Cycler, 7000 System, AB 2720 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
20 Assemblies
Shipping Condition
Room Temperature

Contents & Storage

Contains 20 MicroAmp™ 96-Well Reaction Tube⁄Tray Retainer Sets. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text