2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 403095000
Price (USD)/ Each
201.00
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
201.00
Add to cart
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
Catalog number403095000
Price (USD)/ Each
201.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-bromo-2-methylpropanoyl bromide
Molecular FormulaC4H6Br2O
InChI KeyYOCIJWAHRAJQFT-UHFFFAOYSA-N
SMILESCC(C)(Br)C(Br)=O
Molecular Weight (g/mol)229.90
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5070 to 1.5100 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only