Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Bromophenol Blue

Catalog number: 403140050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromophenol Blue

Catalog number: 403140050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 403140050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115-39-9
IUPAC Name
3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C19H10Br4O5S
InChI Key
UDSAIICHUKSCKT-UHFFFAOYSA-N
SMILES
OC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
Appearance (Color)
Pink-orange to red to purple
Appearance (Form)
Powder
Infrared spectrum
Conforms
Clarity of solution
Passes test
Visual transition interval
from pH 3.0 (yellow) to pH 4.6 (blue)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text