Chemical Identifiers
CAS115-39-9
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H10Br4O5S
InChI KeyUDSAIICHUKSCKT-UHFFFAOYSA-N
SMILESOC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
View more