Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Bromophenol Blue sodium salt, ACS reagent

Catalog number: 403160100
10 g, Each
Thermo Scientific Chemicals

Bromophenol Blue sodium salt, ACS reagent

Catalog number: 403160100
10 g, Each
Quantity
Catalog number: 403160100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34725-61-6
IUPAC Name
sodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular Formula
C19H9Br4NaO5S
InChI Key
KGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES
[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
Appearance (Color)
Dark green to brown-black to blue-black
Appearance (Form)
Powder
Infrared spectrum
Conforms
Clarity of solution
Passes test
Visual transition interval
from pH 3.0 (yellow) to pH 4.6 (blue)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text