Thermo Scientific Chemicals

Bromothymol Blue, Pure, Conform to ACS

Catalog number: 403250100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromothymol Blue, Pure, Conform to ACS

Catalog number: 403250100
10 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 403250100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-59-5
IUPAC Name
3,3-bis[3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C27H28Br2O5S
InChI Key
NUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(=C(C)C(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(Br)=C1C
Appearance (Color)
Light pink to brown to purple
Infrared spectrum
Conforms
Visual transition interval
from pH 6.0 (yellow) to pH 7.6 (blue)
Appearance (Form)
Crystalline powder
Clarity of solution
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text