Thermo Scientific Chemicals

1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 403320250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 403320250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 403320250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2386-60-9
IUPAC Name
butane-1-sulfonyl chloride
Molecular Formula
C4H9ClO2S
InChI Key
WEDIIKBPDQQQJU-UHFFFAOYSA-N
SMILES
CCCCS(Cl)(=O)=O
Appearance (Color)
Colorless to light yellow
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4540 to 1.4570 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text