Thermo Scientific Chemicals

2-Butanone oxime, 99%, Thermo Scientific Chemicals

Catalog number: 403330250
25 g, Each
Thermo Scientific Chemicals

2-Butanone oxime, 99%, Thermo Scientific Chemicals

Catalog number: 403330250
25 g, Each
Quantity
Catalog number: 403330250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-29-7
IUPAC Name
(E)-N-(butan-2-ylidene)hydroxylamine
Molecular Formula
C4H9NO
InChI Key
WHIVNJATOVLWBW-SNAWJCMRSA-N
SMILES
CC\C(C)=N\O
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4410 to 1.4440 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text