Thermo Scientific Chemicals

N-tert-Butylacrylamide, 97%, pure

Catalog number: 403451000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-tert-Butylacrylamide, 97%, pure

Catalog number: 403451000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 403451000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-58-4
IUPAC Name
N-tert-butylprop-2-enamide
Molecular Formula
C7H13NO
InChI Key
XFHJDMUEHUHAJW-UHFFFAOYSA-N
SMILES
CC(C)(C)NC(=O)C=C
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Melting point
126°C to 131°C
Appearance (Form)
Powder and/or chunks
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text