CAB-O-SIL™ M-5, Scintillation Grade
CAB-O-SIL™ M-5, Scintillation Grade
CAB-O-SIL™ M-5, Scintillation Grade
CAB-O-SIL™ M-5, Scintillation Grade
Thermo Scientific Chemicals

CAB-O-SIL™ M-5, Scintillation Grade

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
403731500150 gPlastic Bottle
Catalog number 403731500
Price (USD)
126.65
Special offer
Online exclusive
Ends: 26-Sep-2025
156.00
Save 29.35 (19%)
Each
Add to cart
Quantity:
150 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 26-Sep-2025
156.00
Save 29.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Particle size=<0.02 % 325 Mesh residue
Appearance (Form)Powder
Loss on heating=<1.5 % (105°C, 2 h)
pH3.8 to 4.3 (4 % slurry in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography, For fluorescence/luminescence spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link