Thermo Scientific Chemicals

Calcium carbonate, ACS reagent, chelometric standard, Thermo Scientific Chemicals

Catalog number: 403805000
500 g, Each
Thermo Scientific Chemicals

Calcium carbonate, ACS reagent, chelometric standard, Thermo Scientific Chemicals

Catalog number: 403805000
500 g, Each
Quantity
Catalog number: 403805000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-34-1
IUPAC Name
calcium carbonate
Molecular Formula
CCaO3
InChI Key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES
[Ca++].[O-]C([O-])=O
Potassium (K)
=<0.01 %
Sodium (Na)
=<0.01 %
Strontium (Sr)
=<0.1 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text