Thermo Scientific Chemicals

Calcium Formate, 98%, Pure, Thermo Scientific Chemicals

Catalog number: 403830010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Calcium Formate, 98%, Pure, Thermo Scientific Chemicals

Catalog number: 403830010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 403830010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
544-17-2
IUPAC Name
calcium diformate
Molecular Formula
C2H2CaO4
InChI Key
CBOCVOKPQGJKKJ-UHFFFAOYSA-L
SMILES
[Ca++].[O-]C=O.[O-]C=O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text