Thermo Scientific Chemicals

Calcium hydroxide, 95+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 403850010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium hydroxide, 95+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 403850010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 403850010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1305-62-0
IUPAC Name
calcium dihydroxide
Molecular Formula
CaH2O2
InChI Key
AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Ca++]
Potassium (K)
=<500 ppm
Heavy metals (ICP-OES)
=<30 ppm
Insoluble matter
=<0.03 % (in HCl)
Sulfur compounds (as SO4)
=<0.1 %
Iron (Fe)
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text