Thermo Scientific Chemicals

Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 403880025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 403880025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 403880025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5794-28-5
IUPAC Name
calcium hydrate oxalate
Molecular Formula
C2H2CaO5
InChI Key
LQHWSGSWNOHVHO-UHFFFAOYSA-L
SMILES
O.[Ca++].[O-]C(=O)C([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration
>=97.5 % w/w (as Oxalate) On dry substance
Loss on drying
=<13 % (1 g, 200°C, 3 h)
Insoluble matter
=<0.3 % (in HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text