Calcium propionate, 98+%
Calcium propionate, 98+%
Calcium propionate, 98+%
Calcium propionate, 98+%
Thermo Scientific Chemicals

Calcium propionate, 98+%

CAS: 4075-81-4 | C6H10CaO4 | 186.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 403900010
Price (USD)
52.65
Special offer
Online exclusive
Ends: 26-Sep-2025
65.00
Save 12.35 (19%)
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 26-Sep-2025
65.00
Save 12.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS4075-81-4
IUPAC Namecalcium dipropanoate
Molecular FormulaC6H10CaO4
InChI KeyBCZXFFBUYPCTSJ-UHFFFAOYSA-L
SMILES[Ca++].CCC([O-])=O.CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration Complexometric>=98.0 % (on dry substance)
Loss on drying=<4 % (105°C, 2 Hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link