Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number 404030025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
Molecular Weight (g/mol)12.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Appearance (Form)Powder
Loss on drying=<10 %
Molasses number=<390
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only