Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Thermo Scientific Chemicals

Activated charcoal, NORIT™ CN 1, decolorizing

CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 404042500
Price (USD)
39.90
Each
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
39.90
Each
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Appearance (Form)Powder
pH5.5 to 8
Phosphate (PO4)=<3.5 % (by acid extraction)
Calcium (Ca)=<200 ppm (by acid extraction)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468832Certificate of AnalysisJan 31, 2025404042500, 404045000
A0443910Certificate of AnalysisJan 31, 2025404042500, 404045000
A0463883Certificate of AnalysisJun 19, 2024404042500, 404045000
A0460468Certificate of AnalysisMar 01, 2024404042500, 404045000
A0432421Certificate of AnalysisJan 15, 2024404042500, 404045000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection