Thermo Scientific Chemicals

3,3'-Carbonylbis(7-diethylaminocoumarin), 99%, Thermo Scientific Chemicals

Catalog number: 404050050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,3'-Carbonylbis(7-diethylaminocoumarin), 99%, Thermo Scientific Chemicals

Catalog number: 404050050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 404050050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63226-13-1
IUPAC Name
7-(diethylamino)-3-[7-(diethylamino)-2-oxo-2H-chromene-3-carbonyl]-2H-chromen-2-one
Molecular Formula
C27H28N2O5
InChI Key
SANIRTQDABNCHF-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C2C=C(C(=O)C3=CC4=CC=C(C=C4OC3=O)N(CC)CC)C(=O)OC2=C1
Appearance (Color)
Yellow to orange
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
HPLC
>=98.5 %
UV Lambda max
447 to 451 nm (in toluene)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text