Thermo Scientific Chemicals

5(6)-Carboxyfluorescein

Catalog number: 404100050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

5(6)-Carboxyfluorescein

Catalog number: 404100050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 404100050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72088-94-9
Appearance (Color)
Orange to brown
Infrared spectrum
Conforms
Molar absorption coefficient
>=72000 l/(mol.cm) (at Lamda max 492 nm, in 0.01N NaOH)
Appearance (Form)
Crystalline powder and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text